Anti-MAPT Rabbit Polyclonal Antibody

Toimittaja: Biorbyt
ORB106360-100 ORB106360-20
BIRBORB106360-100EA 0 EUR
BIRBORB106360-100 BIRBORB106360-20
Anti-MAPT Rabbit Polyclonal Antibody
Vasta-aineet
Western Blot: 1:500-1:2000

Type: Primary
Antigen: MAPT (microtubule-associated protein tau)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR