Anti-RPS6KB1 Rabbit Polyclonal Antibody

Toimittaja: Biorbyt
ORB106097-100
BIRBORB106097-100EA 0 EUR
BIRBORB106097-100
Anti-RPS6KB1 Rabbit Polyclonal Antibody
Western Blot: 1:500-1:2000; Immunochemistry: 1:50-1:200

Type: Primary
Antigen: RPS6KB1 (ribosomal protein S6 kinase, 70kDa, polypeptide 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR