Anti-OCIAD2 Rabbit Polyclonal Antibody

Toimittaja: Biorbyt
ORB101509-100 ORB101509-200
BIRBORB101509-100EA 0 EUR
BIRBORB101509-100 BIRBORB101509-200
Anti-OCIAD2 Rabbit Polyclonal Antibody
Vasta-aineet
Western Blot: 1:100-500; ELISA: 1:500-1000; Immunochemistry: 1:100-500; Immunofluorescence: 1:50-200 (based on 0.5 mg/ml)

Type: Primary
Antigen: OCIAD2 (OCIA domain containing 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR