Anti-LDLRAP1 Rabbit Polyclonal Antibody

Toimittaja: Biorbyt
ORB101357-100 ORB101357-200
BIRBORB101357-100EA 0 EUR
BIRBORB101357-100 BIRBORB101357-200
Anti-LDLRAP1 Rabbit Polyclonal Antibody
Vasta-aineet
Western Blot: 1:100-500; ELISA: 1:500-1000; Immunochemistry: 1:100-500; Immunofluorescence: 1:50-200 (based on 0.5 mg/ml)

Type: Primary
Antigen: LDLRAP1 (low density lipoprotein receptor adaptor protein 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR