Anti-ABCF3 Rabbit Polyclonal Antibody

Toimittaja: Biorbyt
ORB100263-100 ORB100263-200
BIRBORB100263-100EA 0 EUR
BIRBORB100263-100 BIRBORB100263-200
Anti-ABCF3 Rabbit Polyclonal Antibody
Vasta-aineet
Western Blot: 1:100-500; ELISA: 1:500-1000; Immunochemistry: 1:100-500; Immunofluorescence: 1:50-200 (based on 0.5 mg/ml)

Type: Primary
Antigen: ABCF3 (ATP-binding cassette, sub-family F (GCN20), member 3)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR