Human skull model on cervical spine, 4 part
Toimittaja: 3B Scientific
1000047
BINH1000047EA 431 EUR
BINH1000047
Human skull model on cervical spine, 4 part
Mallit Ihmisbiologia
This human skull replica is flexibly mounted on a cervical spine. Features also shown are the hindbrain, spinal cord, cervical nerves, vertebral arteries, basilar artery and rear cerebral arteries. Skull can be removed from stand.
  • High quality original human skull cast
  • Skull is handmade from hard, unbreakable plastic
  • Highly accurate representation of the fissures, foramina, processes, sutures etc.
  • Can be disassembled into skull cap, base of skull and mandible
  • Mandible of skull is mounted on a spring to easily demonstrate natural movement
Dimensions: 20More Product Information