Labsit Laboratory Chairs and Stool
Toimittaja: bimos
91232000340353 9123MG01327753 9123MG01327953 9123MG01328053 9123MG01328553 9123MG01340353 91212000327753 91212000327953 91212000328553 91212000340353 9121MG01327753 9121MG01327953 9121MG01328053 9121MG01328553 9121MG01340353 91232000327753 91232000327953 91232000328053 91232000328553 91252000327753 91252000327953 91252000328053 91252000328553 91252000340353 9125MG01327953 9125MG01328053 9125MG01340353 91272000532217 91212000328053 9125MG01327753 9125MG01328553 9125MG01328052 91252000328052 9125E-MG01 9125E-CP01 9125-SP01-3277 9125-SP01-3280 9125-SP01-3403 9125MG01340352 9125E-9801 91252000327752 9122-2000 9125SP01328552 9121MG01327952 9123SP01327952 9123MG01340352 9123MG01327952
132-1274EA 228 EUR
132-1274 132-1267 132-1270 132-1271 132-1268 132-1269 132-1282 132-1285 132-1283 132-1284 132-1277 132-1280 132-1281 132-1278 132-1279 132-1272 132-1275 132-1276 132-1273 132-1292 132-1295 132-1296 132-1293 132-1294 132-1290 132-1291 132-1289 132-1297 132-1286 132-1287 132-1288 BIMO9125MG01328052 BIMO91252000328052 BIMO9125E-MG01 BIMO9125E-CP01 BIMO9125-SP01-3277 BIMO9125-SP01-3280 BIMO9125-SP01-3403 BIMO9125MG01340352 BIMO9125E-9801 BIMO91252000327752 BIMO9122-2000 BIMO9125SP01328552 BIMO9121MG01327952 BIMO9123SP01327952 BIMO9123MG01340352 BIMO9123MG01327952
Labsit Laboratory Chairs and Stool
Tuolit
Thanks to its innovative Flex functions, Labsit chairs are perfectly suited for laboratory work. Labsit offers outstanding comfort, laboratory ergonomics and intuitive operation.
  • The backrest with its 3-D flex function follows the user’s movements
  • Seat upholstery covered with PU foam or artificial leather, soft and comfortable, washable and antibacterial
  • Resistant to all of the usual disinfectants and chemicals in accordance with ISO 2812
  • Minimal joints thanks to 'Hygienic Design' for simple cleaning and disinfection
  • Backrest in plastic and seat shell in a