Flat flange joint, QUICKFIT®
Toimittaja: SciLabware
FG100
BIBBFG100EA 118 EUR
BIBBFG100
Flat flange joint, QUICKFIT®
Sovittimet Sovittimet, NS-hios
PYREX®, borosilicate glass, Quickfit® joints, clear.
  • Excellent chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
Highest quality flat flange joints. Precision ground for leak-free performance.