Distillation Units, Nitrogen Determination, Quickfit®
Toimittaja: SciLabware
FC16B 46MC MC46/SC MC1/F19/SC
BIBBFC16BEA 237 EUR
BIBBFC16B BIBB46MC BIBBMC46/SC BIBBMC1/F19/SC
Distillation Units, Nitrogen Determination, Quickfit®
Tislausosat Tislaussetti
Comprises Markham still body, Liebig condenser and connecting joint clip. When suitable steam is supplied, the steam passes through the sample to promote distillation and when the steam supply is shut off the apparatus automatically sucks back any residue in the vessel.
  • Enables a large number of distillations to be carried out in rapid succession
  • Evaporator vessel enclosed in a steam jacket
Sertifikaatit: Complies with BS1428: Part B2.