Mortars without pestles
Toimittaja: SciLabware
718242 718247 718234
BIBB718242EA 67.2 EUR
BIBB718242 BIBB718247 BIBB718234
Mortars without pestles
Huhmareet ja survimet Huhmareet
Ideal for mixing and grinding powders.
  • Borosilicate glass
  • Low form model
Tilaustiedot: Please order pestles separately.