Distillation Glassware Sets with Ground Glass Joints 14/23, Quickfit®
Toimittaja: SciLabware
27BU
BIBB27BUEA 514 EUR
BIBB27BU
Distillation Glassware Sets with Ground Glass Joints 14/23, Quickfit®
Tislausosat Tislaussetti
PYREX®, Borosilicate glass.
  • Comprises 9 basic items of Quickfit® glassware all with size 14/23 ground-glass joints for compatibility
  • Allows the construction of at least 8 assemblies including steam distillation, reflux with addition and preparation
  • Scale based on starting material quantities up to 30 g
  • Housed in a robust cardboard box with foam rubber liner to provide safe storage and transit
  • Supplied with a CD-ROM which includes practical details of some typical assemblies
The most popular Quickfit® set for schools, colleges and universities.