Showa 7565 Nitrile Gloves
Toimittaja: Showa
7565L 7565M 7565S 7565XL 7565XXL
BEST7565LEA 48.5 EUR
BEST7565L BEST7565M BEST7565S BEST7565XL BEST7565XXL
Showa 7565 Nitrile Gloves
Käsineet
These powder-free disposable gloves are made of 100% nitrile. They provide protection against chemical penetration and projection.
  • Antistatic
  • Chemical-resistant, water-resistant, increased sensitivity
  • Forearm protection
  • Latex, silicone, plasticisers-free
  • Reduce irritation and allergy risks
The single-use glove is chlorinated to enhance comfort and reduce tackiness within the glove. An added textured finish increased grip without reducing fingertip sensitivity.
Typical applications include: Aerospace, electronics, food, pharmaceutical, healthcare,More Product Information