Puhdastilan pyyhkeet, MicroSeal Supersorb™
Toimittaja: BERKSHIRE
MSSS.0909.8 MSSS121212
BERKMSSS.0909.8EA 1880 EUR
BERKMSSS.0909.8 BERKMSSS121212
Puhdastilan pyyhkeet, MicroSeal Supersorb™
Pyyhkeet, liinat ja sienet Puhdastilan pyyhkeet Kuivat puhdastilan pyyhkeet
These wipes are made of 100% continuous filament sealed-edge polyester and designed to minimise and remove cleanroom contamination in a wide range of applications.
  • Ultrasonic sealed edges
  • Highly absorbent and high abrasion resistance
  • Work well in combination with IPA, acetone and other solvents
  • Sterilisable by autoclave and gamma irradiation
Ideal for applications where high concentrations of liquid are in use as well as for cleaning and wiping rough surfaces.
Sertifikaatit: ISO Class 3 – 5, cleanroom Class 1 – 100. EU grade B/C/D.