Puhdastilan pyyhkeet, Durx® 670
Toimittaja: BERKSHIRE
DR670090920 DR670.2309.20 DR670181810 DR670.0707.12 DR670121220 DR670.0404.10 DR670.0812.20 DR670.06D.20 DR670.404010.4
115-0343EA 172 EUR
115-0343 115-0344 BERKDR670181810 BERKDR670.0707.12 BERKDR670121220 115-1784 141-0109 BERKDR670.06D.20 BERKDR670.404010.4
Puhdastilan pyyhkeet, Durx® 670
Pyyhkeet, liinat ja sienet Puhdastilan pyyhkeet Kuivat puhdastilan pyyhkeet
These non woven hydroentangled cellulose polyester blend wipes are ideal for general cleaning and spillages.
  • Low particles, low fibres and no secondary binders
  • Smooth, highly absorbent and durable
  • Work well with isopropanol and other solvents
  • High wet strength
  • Steam autoclavable
Designed for use in manufacturing devices sensitive to micron particle contamination, non volatile residues and ions.
Suitable for use in Class 100, ISO 5, M3.5 cleanrooms.
Väri: valkoinen
Varoitus: Data accurate at time of publishing, latest test results and testMore Product Information