Media bottles, graduated, with screw cap
Toimittaja: BELLCO GLASS
5636-00125 5636-00250 5636-00533 5636-38500
BELC5636-00125EA 199 EUR
BELC5636-00125 BELC5636-00250 BELC5636-00533 BELC5636-38500
Media bottles, graduated, with screw cap
Pullot Väliainepullot
Manufactured from type 1, class B borosilicate glass.
  • White graduations
  • Marking spot
Supplied with a 33-430 nontoxic, rubber-lined black phenolic cap Large opening and sloping shoulders facilitate pouring and cleaning.