Racks for Leighton tissue culture tubes
Toimittaja: BELLCO GLASS
1917-00016
BELC1917-00016EA 1060 EUR
BELC1917-00016
Racks for Leighton tissue culture tubes
Koeputkitelineet Tube Racks
Stainless steel.
  • Spring clip prevents tube from moving in rack
  • Excellent chemical resistance
  • Excellent temperature resistance
For Leighton type culture tubes.