Pipette canisters, round bottom
Toimittaja: BELLCO GLASS
1287-00430 1287-38250 1287-38430
BELC1287-00430EA 486 EUR
BELC1287-00430 BELC1287-38250 BELC1287-38430
Pipette canisters, round bottom
Astiat ja purkit Pipettiastiat
Pipette canister made of borosilicate glass, class A, type I.
  • Manufactured from Type 1, Class A borosilicate glass
  • Components will withstand repeated dry or steam heat sterilisation