Pipette controller, Pipet-Aid
Toimittaja: BELLCO GLASS
1225-85220
BELC1225-85220EA 1250 EUR
BELC1225-85220
Pipette controller, Pipet-Aid
Pipetit ja pipetintäyttäjät Pipetintäyttäjät
Bellco Pipet-Aids are lightweight, compact units that are ideal for use under laminar flow hoods.
  • Rechargeable, self-contained units
  • Two-finger volume control provides precise fluid uptake and dispensing
  • Three independent speed ranges can be set for both fill and dispense rates
Basic Pipet-Aid: 230 V pump with optional tissue culture nose piece and/or air filter.
Portable Pipet-Aid: Rechargeable self-contained unit with removable stand.
Toimitustiedot: Portable pipet-aid is supplied with battery charger and tissue culture nose piece.