Fume hood, moulded, large, SP Bel-Art
Toimittaja: Bel-Art Products, a Part of SP
H500000003
BELAH500000003EA 4230 EUR
BELAH500000003
Fume hood, moulded, large, SP Bel-Art
Vetokaapit ja kotelot Vetolääkekaapit
PE hood, clear acrylic window.
  • Hood has an acrylic panel that slides up and down on the inner tracks
  • When the door is in closed position, a slight opening remains to facilitate airflow
  • Exhaust stack with outside diameter of 150 mm.
Shipping weight: 25,5 kg