Huhmareet, huhmareet jäisten näytteiden jauhamiseen, ilman survinta
Toimittaja: Bel-Art Products, a Part of SP
H372600000 H372600001
BELAH372600000EA 925 EUR
BELAH372600000 BELAH372600001
Huhmareet, huhmareet jäisten näytteiden jauhamiseen, ilman survinta
Huhmareet ja survimet Huhmareet
Liquid nitrogen cooled mortars without pestles, for grinding frozen samples.
  • Insulated handle is removable to save space; 10,2cm long
  • Three suction cup feet stabilise the unit on the workbench
  • Recommended size pestle is any size appropriate for a 275ml mortar (not included)
Grind frozen samples without liquid nitrogen hazards. This 304 stainless steel mortar is suspended by three brackets above a stainless steel bowl which holds liquid nitrogen. The bowl is nested in a heavily insulated blue polyethylene housing.