Drawer organiser trays, SP Bel-Art
Toimittaja: Bel-Art Products, a Part of SP
H186480000 H186550000 H186540000 H186520000
BELAH186480000EA 196 EUR
BELAH186480000 BELAH186550000 BELAH186540000 BELAH186520000
Drawer organiser trays, SP Bel-Art
Haihdutusmaljat/Tarjottimet/Altaat Alustat, säilytyslokerikot
Drawer organiser, made of white polystyrene.
  • Fits into standard laboratory bench drawers
  • Easy to clean
Various configurations available to accommodate thermometers, medication cups, jars, stir bars, vials, and pipettes.