Micro-Tube homogeniser system, Bel-Art ProCulture, SP Bel-Art
Toimittaja: Bel-Art Products, a Part of SP
F650009001 F650000002 F650009002 F650000005 F650000006 F650009006 F651000000
BELAF650009001EA 339 EUR
BELAF650009001 BELAF650000002 BELAF650009002 BELAF650000005 BELAF650000006 BELAF650009006 BELAF651000000
Micro-Tube homogeniser system, Bel-Art ProCulture, SP Bel-Art
Homogenisaattorit Homogenisaattorit, kädessä pidettävät
Hand-held, cordless homogeniser features a simple push in / pull out pestle exchange and is ideal for thorough and faster processing of one to many samples.
  • Pestles and tubes assure tight tolerances and thorough homogenisation and grinding of materials resulting in optimal efficiency of pellet reconstitution or re-suspension
  • Pestles and tubes are chemical and heat resistant; colorant-free to eliminate potential leaching of heavy metals into samples
  • Tubes have 25% thicker bottoms than industry standard to ensure integrity during use
  • Each tube has frosted