Magnetic stirring bars, PTFE, star-shaped, Spinstar®, SP Bel-Art
Toimittaja: Bel-Art Products, a Part of SP
F371460100 F371460250 F371460400
BELAF371460100EA 90.7 EUR
BELAF371460100 BELAF371460250 BELAF371460400
Magnetic stirring bars, PTFE, star-shaped, Spinstar®, SP Bel-Art
Sekoittajat Magneettisauva tai namu
Spinstar® star-shaped stirring bars creates a deep vortex at relatively slow speeds. Bars are an exact fit for the inside diameter of the beaker indicated.