Magnetic stirring bars, PTFE, elliptical, Spinbar®, SP Bel-Art
Toimittaja: Bel-Art Products, a Part of SP
F371300001 F371300112 F371300114 F371300038 F371300034
BELAF371300001EA 26.8 EUR
BELAF371300001 442-9521 BELAF371300114 BELAF371300038 BELAF371300034
Magnetic stirring bars, PTFE, elliptical, Spinbar®, SP Bel-Art
Sekoittajat Magneettisauva tai namu
These PTFE covered egg-shaped magnetic stirring bars have their own pivot point without a separate ring or abrupt change in contour. Designed especially for use in round-bottom vessels, they spin unrestricted by the contours of the vessel.