Drying tubes with tube fittings, SP Bel-Art
Toimittaja: Bel-Art Products, a Part of SP
F199600000 F199610000 F199620000
BELAF199600000EA 68 EUR
BELAF199600000 BELAF199610000 BELAF199620000
Drying tubes with tube fittings, SP Bel-Art
Putket Kuivausputket
Unbreakable polyethylene straight tubes.
  • Good chemical resistance
  • Godd temperature resistance
  • Drying agent no included
Can be filled with desiccant of your choice. Each tube is supplied with two polypropylene fittings which accept 6,4 to 9,5 mm int.Ø tubing.