Contact plate racks, SP Bel-Art
Toimittaja: Bel-Art Products, a Part of SP
F189900000
BELAF189900000EA 194 EUR
BELAF189900000
Contact plate racks, SP Bel-Art
Koeputkitelineet Petrimaljojen säilytystelineet
PP moulded rack, ideal for high volume sampling processes.
  • Transport and incubate contact plates and organise bench top sample processing
  • Racks hold 60 contact plates with covers, configured in six columns of ten plates
  • Interlocking when stacked
  • Large centre divider serves as a handle that does not interfere with samples and provides space for labelling
  • Autoclavable at 121 °C
Some simple assembly required. Dimensions provide stability and conserve bench space: 266×165×178 mm.
Four fully loaded racks can be safely stacked to maximise incubator space.More Product Information