Glassware draining racks, electric, benchtop, Lab-Aire® II, SP Bel-Art
Toimittaja: Bel-Art Products, a Part of SP
F188192304 F188192301
BELAF188192304EA 4660 EUR
BELAF188192304 BELAF188192301
Glassware draining racks, electric, benchtop, Lab-Aire® II, SP Bel-Art
Koeputkitelineet Kuivatustelineet
ABS plastic.
  • Heat selector knob with three temperature settings: high, medium and low
  • Independent thermo-fuse protects unit from overheating
Electric benchtop dryer offer a compact footprint and accelerate drying of labware with warm, circulating air. Electric drying efficiency is enhanced with a replaceable polyurethane foam filter that keeps warmed air dust free.