B.O.D. Bottle Rack, Poxygrid®
Toimittaja: Bel-Art Products, a Part of SP
F169690012
BELAF169690012EA 266 EUR
BELAF169690012
B.O.D. Bottle Rack, Poxygrid®
Koeputkitelineet Pullotelineet
Racks have 12 places (3×4 rows) for 300 ml Biological Oxygen Demand bottles.
  • Epoxy-coated steel wire construction
  • Chemical- and corrosion-resistant
  • Stackable for storage and refrigeration
  • Autoclavable
Made from epoxy-coated steel wire, they are stackable for storage or refrigeration, allowing 16,7 cm between racks. Chemical- and corrosion-resistant, they are autoclavable at 121 °C.