Suppilo, jauheille, kartiomaisella varrella
Toimittaja: Bel-Art Products, a Part of SP
F146810000
BELAF146810000EA 1340 EUR
BELAF146810000
Suppilo, jauheille, kartiomaisella varrella
Suppilot Jauhesuppilot
PP, translucent.
  • Very good chemical resistance
  • Very good temperature resistance
  • Autoclavable at 121 °C
With standard tapered stem.