Pesupullo, turvamerkinnällä, leveä suuaukko, printatulla nimellä
Toimittaja: Bel-Art Products, a Part of SP
F116460628 F116460459 F116460620 F116460622 F116460623 F116460624 F116460638 F116466032 F116461132 F116460621 F116460611 F116460612 F116460614 F116460627 F116460631 F116460639 F116462232 F116462432 F116463832 F116466250 F116460640
215-6233EA 105 EUR
215-6233 BELAF116460459 BELAF116460620 BELAF116460622 BELAF116460623 BELAF116460624 BELAF116460638 BELAF116466032 BELAF116461132 BELAF116460621 BELAF116460611 BELAF116460612 BELAF116460614 BELAF116460627 BELAF116460631 BELAF116460639 BELAF116462232 BELAF116462432 BELAF116463832 BELAF116466250 BELAF116460640
Pesupullo, turvamerkinnällä, leveä suuaukko, printatulla nimellä
Pullot Pesupullot
LDPE, with PP spray cap.
  • Excellent chemical resistance
  • Excellent temperature resistance
  • Wide opening for easy adn safe filling
These bottles include a 53 mm wide PP closure with leakproof design for dispensing most common solvents. Easy-to-read, LDPE solvent-resistant, printed bottles include: Department of Transportation (DOT) and National Fire Protection Association (NFPA) codes and symbols, Chemical Abstract Service (CAS) number, protective equipment symbols and target organ information, making your workplace a safer place. Provides compliance withMore Product Information