Pipette bulbs, Howorka-Ball®
Toimittaja: BEHR
QL9273826 QL9273827
BEHRQL9273826EA 84.2 EUR
BEHRQL9273826 BEHRQL9273827
Pipette bulbs, Howorka-Ball®
Pipetit ja pipetintäyttäjät Pipetintäyttäjät, kumiset
These safety pipette bulbs are suitable for all standard tube diameters, including pipettes, burettes and filter tubes.
  • No rigid connection with tube
  • No valves or side vent on tubes
  • No unstable air pockets
  • Long operational life due to durable components
  • Easily cleaned and steam sterilised if contaminated with liquids