Multi-position heaters, behrotest®
Toimittaja: BEHR
B00217698 B00247310
BEHRB00217698EA 2610 EUR
BEHRB00217698 BEHRB00247310
Multi-position heaters, behrotest®
Lämmittimet Lämpölevyt
Multi-position hotplates with protective metal grills. Maximum plate temperature is 400 °C.
  • Individually adjustable heating controls and indicator lights