GenFind V3 Reagent Kits
Toimittaja: Beckman Coulter
C34880
BECKC34880EA 1370 EUR
BECKC34880
GenFind V3 Reagent Kits
Nukleiinihapporeagenssit Nucleic Acid Purification Kits and Reagents
This prep system kit isolates and purifies genomic DNA (gDNA) from whole blood. The kit is uses Solid Phase Reversible Immobilisation (SPRI) paramagnetic bead-based technology to effectively produce a high recovery of high quality DNA for downstream applications such as PCR.
  • Automated available
Application: Genomic DNA extraction from whole blood.