Male-Female Luer Lock Caps
Toimittaja: BD Medical
TP-MF-O
BDAMTP-MF-OEA 92.9 EUR
BDAMTP-MF-O
Male-Female Luer Lock Caps
Tulpat ja korkit Koeputkitulpat
Caps for Luer lock and Luer slip. Combistoppers for female and male Luer lock.
  • Individual blisters (except for 394080 and 302834 that come in a tray)
DEHP or natural rubber latex are not part of the material formulation.