Peripheral IV cannulae, Insyte™ Autoguard™
Toimittaja: BD Medical
381954
BDAM381954EA 1240 EUR
BDAM381954
Peripheral IV cannulae, Insyte™ Autoguard™
Neulat
Sterile disposable cannulae made of bio materials.
  • Automatic stop of the blood
  • Safety technology reducing by 95% the risks of injuries
  • Apparels are protected and cross contamination is avoided
Suitable for blood controls.