Blood collection needles, BD Vacutainer® Eclipse™
Toimittaja: BD Medical
368609 368609UK 368650 368651
BDAM368609EA 63.4 EUR
BDAM368609 613-3110 BDAM368650 BDAM368651
Blood collection needles, BD Vacutainer® Eclipse™
Ruiskuneulat
The BD Vacutainer® Eclipse™ blood collection needle is a safety-engineered, multi-sample blood collection needle that offers a simple, effective way to collect blood while reducing the possibility of needlestick injuries. It features a safety shield that allows for one-handed activation to cover the needle immediately upon withdrawal from the vein and confirms proper activation with an audible click.
  • Built-in safety shield is aligned with the bevel of the needle
  • The safety shield permanently locks into place, significantly reducing the risk of needlestick