Urine collection tubes and kits, BD Vacutainer®
Toimittaja: BD Medical
364915 365000 368501 364955 364958 368500 364944 368500UK 364915UK 364938UK 364938
BDAM364915EA 113 EUR
BDAM364915 BDAM365000 BDAM368501 BDAM364955 BDAM364958 BDAM368500 BDAM364944 BDAM368500UK BDAM364915UK BDAM364938UK BDAM364938
Urine collection tubes and kits, BD Vacutainer®
Diagnostiset testit Clinical Sample Collection/Preparation Urine Sample Collection/Preparation
This urine collection system is a standardised and hygienic system that can be used right where the sample is taken. It provides the advantages of a closed system that will provide reliable diagnostic results with reduced risk of healthcare worker exposure to hazardous specimens.
  • Made of PET
  • Provided with paper labels
For urinalysis, a wide range of volumes for all patient types, with or without preservative, to be used with collection devices, specimen cups, 24 hour 3 L containers and transfer straws.
For microbiology determinations, a wide rangeMore Product Information