Critical care syringes, BD Vacutainer®
Toimittaja: BD Medical
364356
BDAM364356EA 496 EUR
BDAM364356
Critical care syringes, BD Vacutainer®
Ruiskut Ruiskut, lääketieteellinen käyttö
Critical care syringes can be used to measure arterial blood gas, ionised calcium, ionised magnesium, as well as perform critical care tests.
  • Permits aspiration
Syringes contain calcium balanced lithium heparin spray dried on the sides of the syringe.