Matrigel® Matrix 3-D plates, Corning®
Toimittaja: Corning
356256 356257 356258 356259
BDAA356256EA 1330 EUR
BDAA356256 BDAA356257 BDAA356258 BDAA356259
Matrigel® Matrix 3-D plates, Corning®
Kuoppalevyt Soluviljelylevyt, monikuoppaiset
The Corning® Matrigel® Matrix 3-D plate is a high throughput pre-coated format that contains Matrigel that can be used for drug discovery applications with 3-D cell cultures such as organoids, spheroids or tumoroids.
  • Each lot is verified for intraplate consistency
  • Tested and found negative for the presence of bacteria and fungi
  • Available in 384- and 96-well ready to use formats
  • Easy and convenient as Matrigel is pre-dispensed in each well
  • Formats provide intra-plate consistency
  • Allows for sandwich/overlay and embedded application assays
Order Now
TEKNISET TIEDOT
Environmentally Preferable Low Manufacturing Impact
Ympäristösertifikaatti ISO 14001 - Bedford