Magnetic hotplate stirrers, Cimarec+™
Toimittaja: Thermo Fisher Scientific
SP88854105 SP88857105 SP88857107 SP88857108 SP88850105
BARNSP88854105EA 504 EUR
BARNSP88854105 BARNSP88857105 BARNSP88857107 BARNSP88857108 BARNSP88850105
Magnetic hotplate stirrers, Cimarec+™
Sekoittajat Lämmittävät magneettisekoittajat
These magnetic hotplate stirrers are designed to provide precise stirring control, exceptional safety and temperature performance for routine tasks. They are available in three sizes, providing flexibility from microscale chemistries to production operations. Units feature innovative engineering for slow-speed stirring and strong magnetic coupling. Top plates are available in a choice of ceramic or aluminium and there is a display design that protects the electronics from spills.
  • Easy-to-read LED display for heating and stirring
  • Hot top warning indicator