Cimarec+™ Hotplate
Toimittaja: Thermo Fisher Scientific
HP88857108
BARNHP88857108EA 399 EUR
BARNHP88857108
Cimarec+™ Hotplate
Lämmittimet Lämpölevyt
The Thermo Scientific Cimarec+™ series digital hotplates are designed to provide exceptional safety, and temperature performance for routine protocols
All units offer digital displays of temperature. Temperature is adjustable in 1° increments. The HOT TOP warning system is a standard feature, and is designed to protect users from accidental burns. The HOT TOP feature provides a prominent display when heating surface is above 50 °C, even when unit is shut off (as long as the unit remains plugged in). The Cimarec+™ series also features automatic over-temperature protection, and a raisedMore Product Information