Ultrasonic baths, BactoSonic®
Toimittaja: BANDELIN-ELECTRONIC
3290 3291
BAND3290EA 5120 EUR
BAND3290 BAND3291
Ultrasonic baths, BactoSonic®
Ultraäänihauteet
Bactosonic gently removes the biofilms from implant surfaces.
  • Works with very low ultrasound intensity
  • Biofilm is removed without killing bacteria
  • Compare to standard methods up to 10000 times more bacteria can be detected
Toimitustiedot: Complete set includes ultrasonic bath, wire frame for foil test, TICKOPUR TR-3 concentrate (250 ml), implant boxes, and box trays.