Brilliant cresyl blue ALD, certified by the Biological Stain Commission, Sigma-Aldrich®

Toimittaja: SIGMA ALDRICH MICROSCOPY


M-Clarity™ quality level = MQ200

B5388-10G
B5388-10GEA 240 EUR
B5388-10G
Brilliant cresyl blue ALD, certified by the Biological Stain Commission, Sigma-Aldrich®
Brilliant cresyl blue

Certified as a supravital stain for reticulocytes.

Kaava: C₃₄H₄₂Cl₄N₈O₂Zn
MDL numero: MFCD00148901
CAS-numero: 51716-96-2
EINECS: 279-675-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a5a800920fc11c04f6ca20961be200c5bbfb389a&vid=db763e268b14a2c54715e72d93974ccb04fb2b1c&prid=web&clid=SAPDAM

Order Now

Testitulosten erittely

Dye content ≥60%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR