Indole-5-carbaldehyd ≥98%

Toimittaja: Thermo Fisher Scientific
Varoitus

B25517.03 B25517.06
B25517.03EA 84.7 EUR
B25517.03 B25517.06
Indole-5-carbaldehyd ≥98%
Indole-5-carbaldehyd

Purity: 98% CAS No.: 1196-69-6 Molecular Formula: C9H7NO Formula Weight: 145.16 Melting Point: 96-100° MDL No.: MFCD02093664

Kaava: C₉H₇NO
Sulamispiste: 96…100 °C
Storage Temperature: Huoneenlämpö
MDL numero: MFCD02093664
CAS-numero: 1196-69-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR