Methyl-5-chloro-5-oxovalerate 97%

Toimittaja: Thermo Fisher Scientific
Vaara

B25141.09 B25141.18 B25141.30
B25141.09EA 149 EUR
B25141.09 B25141.18 B25141.30
Methyl-5-chloro-5-oxovalerate 97%
Methyl-5-chloro-5-oxovalerate

Purity: 97% CAS No.: 1501-26-4 Molecular Formula: C6H9ClO3 Formula Weight: 164.59 Storage Temperature: 2°C to 8°C Refractive Index: 1.4470 Boiling Point: 108-110°/15mm UN No.: UN3265 MDL No.: MFCD00000756

Kaava: C₆H₉ClO₃
Kiehumispiste: 108…110 °C (15 mmHg)
MDL numero: MFCD00000756
CAS-numero: 1501-26-4
EINECS: 216-115-6
ADR: 8,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR