Methyl-6-bromo-2-naphthoate ≥98%

Toimittaja: Thermo Fisher Scientific

B25106.06 B25106.14
B25106.06EA 140 EUR
B25106.06 B25106.14
Methyl-6-bromo-2-naphthoate ≥98%
Methyl-6-bromo-2-naphthoate

Purity: 98% CAS No.: 33626-98-1 Molecular Formula: C12H9BrO2 Formula Weight: 265.11 Melting Point: 121-123° Beilstein Reference: 2578943

Kaava: C₁₂H₉BrO₂
Sulamispiste: 121…123 °C
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00100408
CAS-numero: 33626-98-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR