α,α'-Dibromi-m-ksyleeni ≥97%

Toimittaja: Thermo Fisher Scientific
Vaara

B24959.09 B24959.18 B24959.30
B24959.09EA 179 EUR
B24959.09 B24959.18 B24959.30
α,α'-Dibromi-m-ksyleeni ≥97%
α,α'-Dibromi-m-ksyleeni

Purity: 97% CAS No.: 626-15-3 Molecular Formula: C8H8Br2 Formula Weight: 263.96 Melting Point: 74-76° Beilstein Reference: 971085 UN No.: 3261 MDL No.: MFCD00000178

Kaava: C₈H₈Br₂
Kiehumispiste: 135…140 °C (20 mmHg)
Sulamispiste: 74…76 °C
Tiheys: 1,959 g/cm³ (0 °C)
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00000178
CAS-numero: 626-15-3
EINECS: 210-931-6
UN: 3261
ADR: 8,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR