2-(Tributyylistannyyli)pyridiini tech. 80%, Technical Grade

Toimittaja: Thermo Fisher Scientific
Vaara

B24911.03 B24911.06 B24911.14
B24911.03EA 109 EUR
B24911.03 B24911.06 B24911.14
2-(Tributyylistannyyli)pyridiini tech. 80%, Technical Grade
2-(Tributyylistannyyli)pyridiini

Purity: 80% CAS No.: 17997-47-6 Molecular Formula: C17H31NSn Formula Weight: 368.15 Storage Temperature: 2°C to 8°C Refractive Index: 1.5130 Boiling Point: 130-132°/0.8mm Beilstein Reference: 479329 UN No.: 2788 MDL No.: MFCD00052052

Kaava: C₁₇H₃₁NSn
Kiehumispiste: 130…132 °C (0,8 mmHg)
Tiheys: 1,15
Storage Temperature: Jääkaappi
MDL numero: MFCD00052052
CAS-numero: 17997-47-6
UN: 2788
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR