Ethyl-4-phenylpyrrole-3-carboxylate 97%

Toimittaja: Thermo Fisher Scientific
Varoitus

B24877.03 B24877.06 B24877.14
B24877.03EA 110 EUR
B24877.03 B24877.06 B24877.14
Ethyl-4-phenylpyrrole-3-carboxylate 97%
Ethyl-4-phenylpyrrole-3-carboxylate

Purity: 97% CAS No.: 64276-62-6 Molecular Formula: C13H13NO2 Formula Weight: 215.25 Melting Point: 126-128° MDL No.: MFCD03931521

Kaava: C₁₃H₁₃NO₂
Sulamispiste: 126…128 °C
Storage Temperature: Huoneenlämpö
MDL numero: MFCD03931521
CAS-numero: 64276-62-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR