Sodium phosphonoformate hexahydrate ≥98%

Toimittaja: Thermo Fisher Scientific
Varoitus

B24784.03 B24784.06 B24784.14
B24784.03EA 148 EUR
B24784.03 B24784.06 B24784.14
Sodium phosphonoformate hexahydrate ≥98%
Sodium phosphonoformate hexahydrate

Purity: 98+% CAS No.: 34156-56-4 Molecular Formula: Na2O3PCO2Na.6H2O Formula Weight: 300.08 (191.98anhy) MDL No.: MFCD00150176

Kaava: Na₂O₃PCO₂Na·6H₂O
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00150176
CAS-numero: 34156-56-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR